Please login to your online account to display your discounted pricing

Alfa Aesar™ Arg-Gly-Tyr-Val-Tyr-Gln-Gly-Leu

For cellular and molecular biology applications

Specifications

Product Type Arg-Gly-Tyr-Val-Tyr-Gln-Gly-Leu
Molecular Weight 955.08
Storage Requirements -30° to -10°C
Sequence Vesicular stomatitis virus nucleoprotein (52-59)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66441EXD

 
alfa aesar™
J66441EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66441MCR

 
alfa aesar™
J66441MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Arg-Gly-Tyr-Val-Tyr-Gln-Gly-Leu
Molecular Weight 955.08
Storage Requirements -30° to -10°C
Sequence Vesicular stomatitis virus nucleoprotein (52-59)
Format Solid
Common Name VSV-8
Regulatory Status RUO
Purification Method HPLC