Please login to your online account to display your discounted pricing

Alfa Aesar™ Arg-Arg-Leu-Ile-Glu-Asp-Ala-Glu-Tyr(PO_3H_2)-Ala-Ala-Arg-Gly-NH_2

For cellular and molecular biology applications

Specifications

Product Type Arg-Arg-Leu-Ile-Glu-Asp-Ala-Glu-Tyr(PO_3H_2)-Ala-Ala-Arg-Gly-NH_2
Molecular Weight 1598.68
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66462MCR

 
alfa aesar™
J66462MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66462LB0

 
alfa aesar™
J66462LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Arg-Arg-Leu-Ile-Glu-Asp-Ala-Glu-Tyr(PO_3H_2)-Ala-Ala-Arg-Gly-NH_2
Molecular Weight 1598.68
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC