Please login to your online account to display your discounted pricing

Arecaidine propargyl ester tosylate, Tocris


Muscarinic agonist

Quantity 10mg

Manufacturer: r & d systems  RDS038310

 View more versions of this product

Catalog No. 038310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 351.42
Molecular Formula C10H13NO2S·C7H8SO3
Synonym N-Methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate
Chemical Name or Material Arecaidine propargyl ester tosylate
CAS 147202-94-6
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Arecaidine propargyl ester tosylate
Assay Percent Range >98%
Molecular Formula C10H13NO2S·C7H8SO3
Formula Weight 351.42
Synonym N-Methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate
Accessories

Store at Room Temperature