Please login to your online account to display your discounted pricing

Arecaidine propargyl ester tosylate, Tocris

Muscarinic agonist

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material Arecaidine propargyl ester tosylate
Assay Percent Range >98%
Molecular Formula C10H13NO2S·C7H8SO3
Formula Weight 351.42
Synonym N-Methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

038310

 
r & d systems
RDS038310
10mg Each for $121.23

038350

 
r & d systems
RDS038350
50mg Each for $509.13
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 351.42
Chemical Name or Material Arecaidine propargyl ester tosylate
Assay Percent Range >98%
Molecular Formula C10H13NO2S·C7H8SO3
CAS 147202-94-6
Synonym N-Methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature