Please login to your online account to display your discounted pricing

Alfa Aesar™ Arecaidine hydrochloride, 98+%

$44.95 - $112.51

Chemical Identifiers

Chemical Name or Material Arecaidine hydrochloride
CAS 6018-28-6
Molecular Formula C7H11NO2·HCl
Formula Weight 177.63
MDL Number MFCD00051993
Beilstein 3913792
Merck Index 14,777
Synonym 1-Methyl-1,2,5,6-tetrahydronicotinic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0734303

 
alfa aesar™
L0734303
1g Each for $112.51

AAL07343MD

 
alfa aesar™
L07343MD
250mg Each for $44.95
Description & Specifications

Specifications

Formula Weight 177.63
Assay ≥98%
Molecular Formula C7H11NO2·HCl
CAS 6018-28-6
Synonym 1-Methyl-1,2,5,6-tetrahydronicotinic acid hydrochloride
MDL Number MFCD00051993
Beilstein 3913792
Merck Index 14,777
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈260°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number QT2087000
Chemical Name or Material Arecaidine hydrochloride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C