Please login to your online account to display your discounted pricing
Archimedes Balance Set
Determine the mass, volume and density of small solid objects or mineral specimens.
Manufacturer: fisher science education  ABAL6
 View more versions of this product
Catalog No. S96163
  • / Each
  • / Case of 8 Each
Description & Specifications
Specifications
Product Type Archimedes balance set class set of six kit
Includes Graduated cylinder, square inner tube and a density specimen
For Use With (Application) Determine the mass, volume and density of small solid objects or mineral specimens