Archimedes Balance Set
Determine the mass, volume and density of small solid objects or mineral specimens.
$23.65 - $74.25
Specifications
For Use With (Application) Determine the mass, volume and density of small solid objects or mineral specimens
Includes Graduated cylinder, square inner tube and a density specimen
View More Specs
Includes: Graduated cylinder, square inner tube and a density specimen
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S96162 Fisher Science Education
ABAL1
Archimedes balance set demonstration model Each for $23.65
Case of 12 Each for $283.50
S96163 Fisher Science Education
ABAL6
Archimedes balance set class set of six kit Each for $74.25
Case of 8 Each for $594.00
Specifications
Description
Also measures common liquids such as water, seawater, sugar solutions, shampoo, etc.
Description & Specifications
Specifications
Determine the mass, volume and density of small solid objects or mineral specimens
Graduated cylinder, square inner tube and a density specimen