missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Arc H2O™ FR Hi-Vis Rain Jacket, Type R Class 3

Catalog No. 170012069
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170012069 X-Large
170012070 Small
170012071 Medium
170012072 Large
170012073 2X-Large
170012074 3X-Large
170012075 4X-Large
170012076 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170012069 Supplier National Safety Apparel™ Supplier No. R30RL06XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Arc H2O FR HI-Vis rain jacket type R class 3

  • Waterproof welded seams with reinforced stress points to provide durability
  • Batwing pattern creating freedom of movement
  • Contoured collar with tuck away hood
  • Covered zipper and snap closures
  • FR hook and loop adjustable tab closures
  • Back ventilation system with FR mesh for increased airflow
  • Covered D-Ring opening access
  • Two reinforced front pockets with FR hook and loop closure
  • 2 in. silver reflective trim
  • Adjustable cuffs
  • Front storm flap
  • FR polyurethane is durable and flexible that will not become brittle as it ages
  • Made in the USA

Specifications

Size X-Large
Gender Unisex
Material Polyurethane-coated FR Cotton
Color Fluorescent Yellow
Closure Type Zipper
Seam Type Welded
Disposable Reusable
Product Type Hi-vis Rain Jacket
Style Jacket
For Use With (Application) Utilities, Electrical Contractors
Length (English) 30.5 in.
Show More Show Less