Please login to your online account to display your discounted pricing

ARC 239 dihydrochloride, Tocris

α2B antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ARC 239 dihydrochloride
Assay Percent Range >98%
Molecular Formula C24H29N3O3·2HCl
Formula Weight 480.43
Synonym 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

092810

 
r & d systems
092810
10mg Each for $177.37

092850

 
r & d systems
092850
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 480.43
Assay Percent Range >98%
CAS 55974-42-0
Synonym 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride
Molecular Formula C24H29N3O3·2HCl
Chemical Name or Material ARC 239 dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature