missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wells Lamont™ Aramid Fiber Sleeves
These 2 ply Aramid Fiber Sleeves are an ideal solution in providing arm protection and comfort. ANSI A3 rating provides great cut protection, and the material made from para-aramid yarns adds safety with flame-resistant properties. Offer excellent protection against cutting and burn hazards.
$6.35 - $123.06
Specifications
Product Type | Knit Sleeve |
---|---|
Material | Aramid Fiber |
Color | Yellow |
Disposable | Reusable |
Cleaning Procedure | Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution. |
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Style | Construction | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Style | Construction | Price | Quantity | ||
19013437
|
Wells Lamont™
SK14 |
14 in. | 35.5 cm | Standard Sleeve | 2-ply |
Each for $6.35
Dozen of 12 Each for $65.52
|
|
||
19053941
|
Wells Lamont™
SK18T |
Sleeve with Thumb Hole | Aramid Fiber with one track |
Dozen of 12 Each for $101.42
|
|
||||
19037306
|
Wells Lamont™
SK18H |
18 in. | 45.7 cm | Sleeve with Thumb Hole | 2-ply |
Dozen of 12 Each for $123.06
|
|
||
19013439
|
Wells Lamont™
WLCSK16 |
16 in. | 40.6 cm | Standard Sleeve | 2-ply |
Each for $6.76
|
|
||
Description
- 2-ply knit sleeve
- Flame resistant
- Available with or without a thumb hole
- ANSI Level 2 Heat Rating
- ANSI Level 4 Flame Rating
- ANSI Cut Level A3
- Made in the USA
Specifications
Knit Sleeve | |
Yellow | |
Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution. |
Aramid Fiber | |
Reusable |