Wells Lamont™ Aramid Fiber Sleeves
Manufacturer:  Wells Lamont™ SK14KCL
 View more versions of this product
Catalog No. 29-636-1640
Description
Description
Specifications
Specifications
Yellow
14 in.
Dupont™ Kevlar™ aramid fibers
1-ply outer Dupont™ Kevlar™ aramid fiber, 1 ply inner Cotton
35.5 cm
100% Dupont™ Kevlar™ aramid fibers with cotton lining