Please login to your online account to display your discounted pricing

Arachidonic acid, 90 to 95%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0210026991

 View more versions of this product

Catalog No. ICN10026991

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 0.922g/mL
Quantity 1g
Chemical Name or Material Arachidonic acid
CAS 506-32-1
Physical Form Clear colorless liquid
Assay Percent Range ≥90%
Molecular Formula C20H32O2
Formula Weight 304.5g/mol
MDL Number MFCD00004417
Synonym Eicosa-5Z, 8Z, 11Z
Storage Note 1 Protect from light.
Recommended Storage Store at 0°C.

Specifications

Chemical Name or Material Arachidonic acid
CAS 506-32-1
Physical Form Clear colorless liquid
Assay Percent Range ≥90%
Molecular Formula C20H32O2
Formula Weight 304.5g/mol
MDL Number MFCD00004417
Synonym Eicosa-5Z, 8Z, 11Z
Accessories

Store at 0°C.

Protect from light.