Please login to your online account to display your discounted pricing

Arachidonic acid, 90 to 95%, MP Biomedicals™

$238.26 - $408.78

Chemical Identifiers

Chemical Name or Material Arachidonic acid
CAS 506-32-1
Physical Form Clear colorless liquid
Assay Percent Range ≥90%
Molecular Formula C20H32O2
Formula Weight 304.5g/mol
MDL Number MFCD00004417
Synonym Eicosa-5Z, 8Z, 11Z
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10026990

 
mp biomedicals
0210026990
500mg Each for $238.26

ICN10026991

 
mp biomedicals
0210026991
1g Each for $408.78
Description & Specifications

Specifications

Density 0.922g/mL
Quantity 500mg
Chemical Name or Material Arachidonic acid
CAS 506-32-1
Physical Form Clear colorless liquid
Assay Percent Range ≥90%
Molecular Formula C20H32O2
Formula Weight 304.5g/mol
MDL Number MFCD00004417
Synonym Eicosa-5Z, 8Z, 11Z
Storage Note 1 Protect from light.
Recommended Storage Store at 0°C.
Safety and Handling

Store at 0°C.

Protect from light.