Please login to your online account to display your discounted pricing

AR-M 1000390 hydrochloride, Tocris™

Low-internalizing δ opioid receptor agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material AR-M 1000390 hydrochloride
Assay Percent Range >97%
Molecular Formula C23H28N2O·HCl
Formula Weight 384.94
Synonym N, N-Diethyl-4-(phenyl-4-piperidinylidenemethyl)-benzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

433510

 
r & d systems
RDS433510
10mg Each for $214.12

433550

 
r & d systems
RDS433550
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 384.94
Quantity 10mg
Recommended Storage Store at -20°C
Assay Percent Range >97%
Chemical Name or Material AR-M 1000390 hydrochloride
Synonym N, N-Diethyl-4-(phenyl-4-piperidinylidenemethyl)-benzamide hydrochloride
Molecular Formula C23H28N2O·HCl
CAS 209808-47-9
Safety and Handling

Store at -20°C