Please login to your online account to display your discounted pricing

AR-C 102222, Tocris™

Selective iNOS inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material AR-C 102222
Assay Percent Range >98%
Molecular Formula C19H16F2N6O·HCl
Formula Weight 418.83
Synonym 5-[(4'-Amino-5', 8'-difluorospiro[piperidine-4, 2'(1'H)-quinaxolin]-1-yl)carbonyl]-2-pyridinecarbonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

396910

 
r & d systems
396910
10mg Each for $290.95

396950

 
r & d systems
396950
50mg Each for $1,205.80
Description & Specifications

Specifications

Formula Weight 418.83
Quantity 10mg
Chemical Name or Material AR-C 102222
Assay Percent Range >98%
Molecular Formula C19H16F2N6O·HCl
CAS 253771-21-0
Synonym 5-[(4'-Amino-5', 8'-difluorospiro[piperidine-4, 2'(1'H)-quinaxolin]-1-yl)carbonyl]-2-pyridinecarbonitrile hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C