Please login to your online account to display your discounted pricing

AR-A 014418, Tocris™

Selective GSK-3 inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material AR-A 014418
Assay Percent Range >99%
Molecular Formula C12H12N4O4S
Formula Weight 308.31
Synonym N-[(4-Methoxyphenyl)methyl]-N'-(5-nitro-2-thiazolyl)urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

396610

 
r & d systems
RDS396610
10mg Each for $218.74

396650

 
r & d systems
RDS396650
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 308.31
Quantity 10mg
Chemical Name or Material AR-A 014418
Assay Percent Range >99%
Molecular Formula C12H12N4O4S
CAS 487021-52-3
Synonym N-[(4-Methoxyphenyl)methyl]-N'-(5-nitro-2-thiazolyl)urea
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C