Please login to your online account to display your discounted pricing

Alfa Aesar™ Aqueous dispersant for multi-walled carbon nanotubes

For dispersing carbon nanotubes

$74.45 - $74.45

Chemical Identifiers

Chemical Name or Material Aqueous dispersant for multi-walled carbon nanotubes
MDL Number MFCD04040667
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44276AB

  SDS
alfa aesar™
44276AB
10mL Each for $74.45
Description & Specifications

Specifications

Quantity 10mL
Physical Form Liquid
Chemical Name or Material Aqueous dispersant for multi-walled carbon nanotubes
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
TSCA TSCA
MDL Number MFCD04040667
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.