Please login to your online account to display your discounted pricing

AQUASTAR™ Titrant 5, EMD Millipore™


Quantity 1L
Packaging 38/430 Poly Coated Glass Bottle

Manufacturer: emd millipore corporation  1880101038

 View more versions of this product

Catalog No. M1880101038

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Color Red-brown
UN Number 1230
Physical Form Liquid
Chemical Name or Material AQUASTAR Titrant 5
CAS 7553-56-2
CAS Min % 50.0
Packaging 38/430 Poly Coated Glass Bottle
CAS 67-56-1
CAS Min % 10.0

Specifications

Chemical Name or Material AQUASTAR Titrant 5
CAS 67-56-1
CAS Min % 50.0
CAS 7553-56-2
CAS Min % 10.0
Physical Form Liquid