Please login to your online account to display your discounted pricing

AQUASTAR™ Titrant 5, EMD Millipore™

$527.97 - $924.03

Chemical Identifiers

Chemical Name or Material AQUASTAR Titrant 5
CAS 67-56-1,7553-56-2
CAS Min % 50.0
CAS 7553-56-2
CAS Min % 10.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1880101038

 
emd millipore corporation
1880101038
38/430 Poly Coated Glass Bottle 1L Each for $527.97

M1880102538

 
emd millipore corporation
1880102538
38/430 Poly Coated Glass Bottle 2.5L Each for $924.03
Description & Specifications

Specifications

Quantity 1L
Color Red-brown
UN Number 1230
Physical Form Liquid
Chemical Name or Material AQUASTAR Titrant 5
CAS 7553-56-2
CAS Min % 50.0
Packaging 38/430 Poly Coated Glass Bottle
CAS 67-56-1
CAS Min % 10.0