Please login to your online account to display your discounted pricing

AQUASTAR™ Titrant 5, EMD Millipore™


Quantity 2.5L
Packaging 38/430 Poly Coated Glass Bottle

Manufacturer: emd millipore corporation  1880102538

 View more versions of this product

Catalog No. M1880102538

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 2.5L
Flash Point <21°C (<69.8°F)
CAS 67-56-1,7553-56-2
UN Number 1230
Density 0.850 to 0.860g/cm3
Color Red-brown
Physical Form Liquid
Chemical Name or Material AQUASTAR Titrant 5
CAS Min % 50.0
CAS 7553-56-2
CAS Min % 10.0
Packaging 38/430 Poly Coated Glass Bottle

Specifications

Chemical Name or Material AQUASTAR Titrant 5
CAS 67-56-1,7553-56-2
CAS Min % 50.0
CAS 7553-56-2
CAS Min % 10.0
Physical Form Liquid