Please login to your online account to display your discounted pricing

AQUASTAR™ CombiTitrant 5, EMD Millipore™

$954.57 - $954.57

Chemical Identifiers

Chemical Name or Material AQUASTAR CombiTitrant 5
CAS 111-90-0
CAS Min % 50.0
CAS Max % 75.0
CAS 7553-56-2
CAS Min % 10.0
CAS Max % 20.0
CAS 288-32-4
CAS Min % 10.0
CAS Max % 20.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1880052538

 
emd millipore corporation
1880052538
38/430 Poly coated glass bottle 2.5L Each for $954.57
Description & Specifications

Specifications

Quantity 2.5L
Color Red-brown
UN Number 1760
Physical Form Liquid
CAS 7446-09-5
CAS Max % 20.0
CAS Max % 75.0
Packaging 38/430 Poly coated glass bottle
CAS Min % 10.0
CAS 111-90-0
Chemical Name or Material AQUASTAR CombiTitrant 5
CAS Max % 10.0
CAS Min % 50.0
CAS 7553-56-2
CAS Max % 20.0
CAS 288-32-4
CAS Min % 10.0
CAS Min % 0.0