Please login to your online account to display your discounted pricing

AQUASTAR™ CombiTitrant 5, EMD Millipore™


Quantity 2.5L
Packaging 38/430 Poly coated glass bottle

Manufacturer: emd millipore corporation  1880052538

 View more versions of this product

Catalog No. M1880052538

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 2.5L
Color Red-brown
UN Number 1760
Physical Form Liquid
CAS 7446-09-5
CAS Max % 20.0
CAS Max % 75.0
Packaging 38/430 Poly coated glass bottle
CAS Min % 10.0
CAS 111-90-0
Chemical Name or Material AQUASTAR CombiTitrant 5
CAS Max % 10.0
CAS Min % 50.0
CAS 7553-56-2
CAS Max % 20.0
CAS 288-32-4
CAS Min % 10.0
CAS Min % 0.0

Specifications

Chemical Name or Material AQUASTAR CombiTitrant 5
CAS 111-90-0
CAS Min % 50.0
CAS Max % 75.0
CAS 7553-56-2
CAS Min % 10.0
CAS Max % 20.0
CAS 288-32-4
CAS Min % 10.0
CAS Max % 20.0
CAS 7446-09-5
CAS Min % 0.0
CAS Max % 10.0
Physical Form Liquid