Please login to your online account to display your discounted pricing

AquaStar Anhydrous Methanol, EMD Millipore


Quantity 1L
Packaging Amber Glass

Manufacturer: emd millipore corporation  AX1699M6

 View more versions of this product

Catalog No. MAX1699M6

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 32.04
Quantity 1L
Color Colorless to pale yellow
DOT Information DOT Class 3, : Flammable Liquid, Toxic
Chemical Name or Material AQUASTAR™ Anhydrous Methanol
UN Number 1230
Physical Form Liquid
Name Note AQUASTAR™
Packaging Amber Glass
Recommended Storage Room Temperature
CAS 67-56-1
Linear Formula CH3OH

Specifications

Chemical Name or Material AQUASTAR™ Anhydrous Methanol
Name Note AQUASTAR™
CAS 67-56-1
Physical Form Liquid
Linear Formula CH3OH
Formula Weight 32.04

  • Methanol for Volumetric Karl Fischer Titrations.
  • For moisture determination. Assay >99.8%. Max. 0.01% H&subscript_2;O content.

Accessories

Room Temperature