Please login to your online account to display your discounted pricing

AquaStar Anhydrous Methanol, EMD Millipore

$172.57 - $172.57

Chemical Identifiers

Chemical Name or Material AQUASTAR™ Anhydrous Methanol
Name Note AQUASTAR™
CAS 67-56-1
Physical Form Liquid
Linear Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MAX1699M6

 
emd millipore corporation
AX1699M6
Amber Glass 1L Each for $172.57
Description & Specifications

Specifications

Formula Weight 32.04
Quantity 1L
Color Colorless to pale yellow
DOT Information DOT Class 3, : Flammable Liquid, Toxic
Chemical Name or Material AQUASTAR™ Anhydrous Methanol
UN Number 1230
Physical Form Liquid
Name Note AQUASTAR™
Packaging Amber Glass
Recommended Storage Room Temperature
CAS 67-56-1
Linear Formula CH3OH
Safety and Handling

Room Temperature