Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical

For two-component system. Water equivalent: 5.0mg H2O/mL

$65.87 - $65.87

Chemical Identifiers

CAS 67-56-1
ChemAlert Storage Symbol Red
Chemical Name or Material Aqualine™ Titrant 5
Cleanup Symbol Solvent
Boiling Point 64°C
Color Clear colorless
Physical Form Liquid
DOT Information DOT Class 3, : Flammable Liquid
Equivalence 5.0mg of water/mL min.
Health Hazard 2 DANGER!
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

K2200-1

 
fisher scientific company
K220015
Amber glass bottle 1L Each for $65.87
Description & Specifications

Specifications

Boiling Point 64°C
Color Clear colorless
Vapor Density > 1.0
Physical Form Liquid
MDL Number 132730
Packaging Amber glass bottle
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Purity or Free of Statement Pyridine free
Chemical Name or Material Aqualine™ Titrant 5
Cleanup Symbol Solvent
Name Note Karl Fischer Reagent
CAS 67-56-1
ChemAlert Storage Symbol Red
UN Number 1992
Equivalence 5.0mg of water/mL min.
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Poison, may be fatal or cause blindness if swallowed. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:3
Instability:0
Safety and Handling
 

Emergency Overview
Poison, may be fatal or cause blindness if swallowed. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:3
Instability:0

DANGER!