Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical


Quantity 1L
Packaging Amber glass bottle

Manufacturer: fisher scientific company  AL21101

 View more versions of this product

Catalog No. AL-21101

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

MDL Number 81734
Packaging Amber glass bottle
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Purity or Free of Statement Pyridine free
CAS 67-56-1
Chemical Name or Material Aqualine™ Solvent CM
CAS 7446-09-5
Use Test Pass Test
Cleanup Symbol Solvent
Name Note Karl Fischer Reagent
ChemAlert Storage Symbol Red
CAS 67-66-3
CAS 288-32-4

Specifications

CAS 67-56-1
CAS 67-66-3
CAS 288-32-4
CAS 7446-09-5
ChemAlert Storage Symbol Red
Chemical Name or Material Aqualine™ Solvent CM
Cleanup Symbol Solvent
Physical Form Liquid
DOT Information DOT Class 3, : Flammable Liquid
MDL Number 81734
Name Note Karl Fischer Reagent
Packaging Amber glass bottle
Purity or Free of Statement Pyridine free
Use Test Pass Test
Accessories
   

SDS & Safety Documents