Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical


Quantity 4L
Packaging Amber Glass

Manufacturer: thermo fisher scientific  AL2300R4

 View more versions of this product

Catalog No. AL2300R-4

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4L
DOT Information DOT Class 6.1, : Poison
Physical Form Liquid
Color Clear Colorless
Chemical Name or Material Aqualine™ Matrix K
Cleanup Symbol Solvent
MDL Number MFCD00006998
CAS 67-66-3
ChemAlert Storage Symbol Blue
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. May cause central nervous system effects. Possible risk of harm to the unborn child. May form explosive peroxides. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:1
Packaging Amber Glass
CAS 111-76-2
CAS 5683-33-0
Use Test Pass test
Name Note Karl Fischer Reagent
CAS 107-21-1

Specifications

Physical Form Liquid

  • Matrix to be used in conjunction with Complete 5K (AL2250R) for high water content analysis of aldehydes and ketones

Accessories

Accessories for Catalog Number: AL2300R4

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Causes burns by all exposure routes. May cause central nervous system effects. Possible risk of harm to the unborn child. May form explosive peroxides. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:1

DANGER!

 

SDS & Safety Documents