missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Supplier: Fisher Chemical K2300R15
Description
- Matrix to be used in conjunction with Complete 5K (AL2250R-1) for high water content analysis of aldehydes and ketones
Specifications
Colorless | |
Aqualine™ Matrix K | |
107-21-1,111-76-2,5683-33-0 | |
Liquid | |
> 1.0 |
1 L | |
Karl Fischer Reagent | |
Amber glass bottle | |
Pass test | |
81734 |