Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical


Quantity 1L
Packaging Amber glass bottle

Manufacturer: fisher scientific company  K2250R15

 View more versions of this product

Catalog No. K2250-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless
MDL Number 81734
Packaging Amber glass bottle
Physical Form Liquid
Quantity 1L
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. May cause central nervous system effects. Possible risk of harm to the unborn child. May form explosive peroxides. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:1
ChemAlert Storage Symbol Gray
Cleanup Symbol Solvent
CAS 7553-56-2
CAS 5683-33-0
Equivalence Report mg of water/mL
Health Hazard 2 DANGER!
Name Note Karl Fischer Reagent
CAS 7446-09-5
CAS 111-90-0
Chemical Name or Material Aqualine™ Complete 5K

Specifications

Physical Form Liquid

  • Single-component volumetric reagent for high water content analysis of aldehydes and ketones
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free

Accessories
 

Emergency Overview
Causes burns by all exposure routes. May cause central nervous system effects. Possible risk of harm to the unborn child. May form explosive peroxides. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:1

DANGER!

 

SDS & Safety Documents