Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical


Single-component reagent; Pyridine free

Quantity 1L
Packaging Amber glass bottle

Manufacturer: fisher scientific company  K200015

 View more versions of this product

Catalog No. K2000-1

  • / Each
 

Description & Specifications

Chemical Identifiers

MDL Number 81734
Packaging Amber glass bottle
Quantity 1L
Physical Form Liquid
Purity or Free of Statement Pyridine free
Color Brown
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation. Possible risk of harm to the unborn child.
NFPA
Health:
Health Hazard 2 DANGER!
Name Note Karl Fischer Reagent
Chemical Name or Material Aqualine™ Complete 5
CAS 111-90-0
Cleanup Symbol Solvent
CAS 7553-09-5
CAS 7446-09-5
CAS 288-32-4
Equivalence 5.2mg of water/mL min.
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:DANGER!

Specifications

CAS 111-90-0
CAS 288-32-4
CAS 7446-09-5
CAS 7553-09-5
ChemAlert Storage Symbol Gray
Chemical Name or Material Aqualine™ Complete 5
Cleanup Symbol Solvent
Color Brown
Physical Form Liquid
Equivalence 5.2mg of water/mL min.
Health Hazard 1 Signal Word:DANGER!
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation. Possible risk of harm to the unborn child.
NFPA
Health:
Name Note Karl Fischer Reagent
Purity or Free of Statement Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free

Accessories
 

Signal Word:DANGER!

Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation. Possible risk of harm to the unborn child.
NFPA
Health:

DANGER!

 

SDS & Safety Documents