missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Single-component reagent; Pyridine free

$96.36 - $227.72

Specifications

CAS 111-90-0,288-32-4,7446-09-5,7553-09-5
Chemical Name or Material Aqualine™ Complete 5
Color Brown
Physical Form Liquid
Equivalence 5.2mg of water/mL min.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
K2000-1
View Documents
Fisher Chemical
K200015
1L
Each for $96.36
 
K2000-212
Accessories View Documents
Fisher Chemical
K200017
2.5L
Each for $227.72
 
Description

Description

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
Specifications

Specifications

111-90-0,288-32-4,7446-09-5,7553-09-5
Brown
5.2mg of water/mL min.
Pyridine free
Aqualine™ Complete 5
Liquid
Karl Fischer Reagent
SDS