Please login to your online account to display your discounted pricing

Karl Fischer Aqualine™ Complete 2, for Karl Fischer Titration By Volumetry, Fisher Chemical


Single-component reagent; Pyridine free

Quantity 1L
Packaging Amber Glass

Manufacturer: fisher scientific company  K195015

 View more versions of this product

Catalog No. K1950-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 1L
Molecular Formula Solution
Purity or Free of Statement Pyridine free
Packaging Amber Glass
Equivalence 2.2mg of water/mL min.
CAS 7553-56-2
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Irritating to skin. Irritating to eyes. Possible risk of harm to the unborn child.
NFPA
Health:
CAS 111-90-0
Chemical Name or Material Aqualine™ Complete 2
Cleanup Symbol Solvent
Health Hazard 1 Signal Word:DANGER!
MDL Number MFCD00081734
CAS 288-32-4
Name Note Karl Fischer Reagent
CAS 7446-09-5
ChemAlert Storage Symbol Gray

Specifications

Physical Form Liquid
Accessories
 

Signal Word:DANGER!

Emergency Overview
Irritating to skin. Irritating to eyes. Possible risk of harm to the unborn child.
NFPA
Health:

DANGER!

 

SDS & Safety Documents