missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AQ-RA 741, Tocris

Selective, high affinity M2 antagonist

$192.67 - $812.12

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C27H37N5O2
Formula Weight 463.62
Synonym 11-[[4-[4-(Diethylamino)butyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
Chemical Name or Material AQ-RA 741
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
229250 R & D Systems
229250
50mg Each for $812.12
229210 R & D Systems
229210
10mg Each for $192.67
Specifications Description & Specifications

Chemical Identifiers

>99%
463.62
AQ-RA 741
C27H37N5O2
11-[[4-[4-(Diethylamino)butyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one

Specifications

123548-16-3
50mg
463.62
AQ-RA 741
>99%
C27H37N5O2
11-[[4-[4-(Diethylamino)butyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one

Safety and Handling

Recommended Storage : Store at Room Temperature