missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Apron Rail

Fisherbrand™ Apron Rails are used to span knee space areas between two cabinets.

Manufacturer:  Fisherbrand™ FJPTBR3602

 View more versions of this product

Catalog No. 06000034



Description

Description

  • Height of apron rail is 2 in.
  • Can be cut to fit in the field

Compliance

SEFA 8

Specifications

Specifications

White
5.1cm
5.1cm
Steel
91.4cm
2 in.
2 in.
Attachment hardware
36 in.
Apron Rail
Documents