Please login to your online account to display your discounted pricing

Alfa Aesar™ Apramycin sulfate, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Apramycin sulfate
CAS 65710-07-8
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD06200257
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67407AC

  SDS
alfa aesar™
J67407AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67407AD

  SDS
alfa aesar™
J67407AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 1 Danger
Physical Form Liquid
MDL Number MFCD06200257
Chemical Name or Material Apramycin sulfate
CAS 65710-07-8
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Storage Note 1 -30° to -10°C
Name Note 50mg/mL in distilled water, sterile-filtered
Quantity 25mL
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Apramycin sulfate is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.

-30° to -10°C