Please login to your online account to display your discounted pricing

Alfa Aesar™ Apramycin sulfate, 50mg/mL in distilled water, sterile-filtered


Quantity 50mL

Manufacturer: alfa aesar™  J67407AD

 View more versions of this product

Catalog No. AAJ67407AD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Apramycin sulfate
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 65710-07-8
MDL Number MFCD06200257
Name Note 50mg/mL in distilled water, sterile-filtered
Storage Note 1 -30° to -10°C
Health Hazard 1 Danger
Physical Form Liquid
Quantity 50mL
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.

Specifications

Chemical Name or Material Apramycin sulfate
CAS 65710-07-8
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD06200257

Applications
Apramycin sulfate is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Accessories

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.

-30° to -10°C

SDS & Safety Documents