Applied Biosystems™ TaqMan™ PreAmp Master Mix Kit DFS Item
Amplifies with no bias - high correlation between amplified and unamplified cDNA
$1,766.10 - $1,766.10
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes: Contains 2X TaqMan PreAmp Master Mix (1mL), TaqMan Gene Expression Master Mix (5mL), and TaqMan PreAmp Master Mix Kit Quick Reference Card.
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
4384267 Applied Biosystems™
LS4384267
Each for $1,766.10
Specifications
Description
Applied Biosystems™ TaqMan™ PreAmp Master Mix Kit contains TaqMan™ PreAmp Master Mix and TaqMan™ Gene Expression Master Mix. Two work hand in hand to provide a seamless workflow for preamplification and quantitation of mRNA.
  • Amplifies cDNA targets equally without introducing bias
  • Analyzes mRNA from any precious sample such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE)
  • Stretches as 1 ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TAqMan Gene Expression Assays
  • Analyzes up to 100 gene expression targets with minimal hands-on time
Drug Discovery and Development, Gene Expression Analysis and Genotyping, PCR and Real-Time PCR, PCR-Based Drug Discovery Assays, RNAi, Epigenetics and Non-Coding RNA Research, Real Time PCR (qPCR), Real Time PCR-Based Gene Expression Profiling, Real Time PCR-Based miRNA Analysis, Target and Lead Identification and Validation, miRNA and Non-Coding RNA Analysis