Applied Biosystems™ GeneAmp™ High Fidelity PCR System DFS Item
Amplify DNA fragments up to 10kb
$155.40 - $758.00
Specifications
Fidelity (vs. Taq) Fidelity (vs. Taq): 3X
Storage Requirements Store at -20°C.
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes: Contains GeneAmp High Fidelity Enzyme Mix, 10X PCR Buffer with MgCl2, 10X PCR Buffer without MgCl2, and MgCl2 solution. Sufficient for 40/200 PCR amplifications, 50μL each.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
4328216 Life Technologies
LS4328216
40 reactions Each for $155.40
4328217 Life Technologies
LS4328217
200 reactions Each for $758.00
Specifications
Description
The Applied Biosystems GeneAmp High Fidelity PCR System is designed for accuracy and increased PCR yield.
  • AmpliTaq™ DNA Polymerase-based enzyme blend is formulated for increased fidelity and yield of PCR products
  • Proofreading activity enables amplification of DNA fragments up to 10 kb
  • Two-buffer system provides versatility
  • Fidelity (vs. Taq): 3X
High Fidelity
The GeneAmp High Fidelity PCR System contains an enzyme blend of AmpliTaq DNA Polymerase and a new thermo stable proofreading enzyme. This proprietary blend increases the fidelity of PCR 3-fold over Taq DNA Polymerase. The careful blending of these enzymes prevents truncated products resulting from misincorporated nucleotides, and limits primer degradation by the 3' to 5' exonuclease activity of the proofreader. The increased sensitivity of the unique GeneAmp High Fidelity Enzyme Mix enables detection down to 1 ng of complex templates such as human genomic DNA.
Flexible Kit
The system is supplied with buffers specifically designed to enhance fidelity, sensitivity, and yield of PCR experiments. Two versions of the buffer are included for flexibility. For ease of use, one version contains a final MgCl2 concentration of 1.5 mM. The second buffer does not contain MgCl2. A separate MgCl2 solution is also provided for optimizing MgCl2 concentration. The system allows for amplification of products ranging from a hundred bases up to 10 kb from human genomic DNA. It can be used with templates from various sources such as mammalian genomic DNA, artificial chromosomes, plasmids, and cDNA.
See user's manual or package insert for limited label license and trademark information.
High Fidelity PCR, Long Fragment PCR, PCR, PCR and Real-Time PCR
Description & Specifications
Specifications
Fidelity (vs. Taq): 3X
Store at -20°C.