missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ GeneAmp™ 10X PCR Gold Buffer & MgCl2 DFS Item

Provides flexible, efficient activation of AmpliTaq Gold DNA Polymerase for highly specific, robust PCR amplification

$36.38 - $273.00

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR
Overhang 3'-A
Size (Final Product) 5kb or less
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
4306894 Life Technologies
LS4306894
1 x 1.5mL each of GeneAmp™ 10X PCR Gold Buffer and 25mM MgCl2 1 x 1.5mL Each for $36.38
4379874 Life Technologies
LS4379874
2 x 75mL each of GeneAmp™ 10X PCR Gold Buffer and 25mM MgCl2 2 x 75mL Each for $273.00
Specifications

Description

When diluted 1:10, GeneAmp10X PCR Gold Buffer provides the optimal pH and ionic strength for PCR activation and amplification with AmpliTaq Gold DNA Polymerase, ensuring maximum performance.

  • Available in several convenient sizes
  • Both magnesium ion free and gelatin free
  • A separate MgCl2 solution is provided
  • When used together, GeneAmp 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template

See user's manual or package insert for limited label license and trademark information.

Hot Start PCR, PCR, PCR and Real-Time PCR

Order Info

Shipping condition: Dry Ice

Description & Specifications

Specifications

Hot Start PCR, PCR, PCR and Real-Time PCR
3'-A
5kb or less