Please login to your online account to display your discounted pricing

Applied Biosystems™ Centri-Spin™ Spin Columns DFS Item

Process samples faster than ultrafiltration columns

$122.00 - $270.00

Specifications

Format Spin Column
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostics procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

4367828

DFS Item
 
life technologies
LS4367828
50 columns Each for $270.00

4367827

DFS Item
 
life technologies
LS4367827
20 columns Each for $122.00

4367830

DFS Item
 
life technologies
LS4367830
50 columns Each for $270.00

4367832

DFS Item
 
life technologies
LS4367832
50 columns Each for $270.00
Description & Specifications

Specifications

Format Spin Column

  • Centri-Spin™ columns provide excellent recovery of RNA and DNA fragments for a variety of purification applications. Use Centri-Spin columns to remove primers, probes, and dNTPs from DNA, RNA, PCR and end-labeled products
  • Rapid and efficient separations
  • Permits user to determine buffer composition
  • Convenient 20-50μL sample size
  • Dry gel format
  • Room Temperature Storage
  • Microcentrifuge format
  • Excellent Recovery
  • Centri-Spin columns provide excellent recovery of DNA fragments with sizes greater than 10, 20 or 40 base pairs (depending on kit), while removing >98% of salts, NTP's and other low-molecular-weight impurities. Proteins, peptides, and protein conjugates >5kD can also be efficiently separated/desalted from unwanted low-molecular-weight impurities
  • Designed for Performance
  • The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Spin consists of a specially fritted microfuge tube, dry gel, a wash tube and a sample collection tube, all specifically designed for this purpose
  • Versatile
  • The column gel can be hydrated with either reagent-grade water or a suitable buffer, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. After the sample is applied, spin the column again to remove the low-molecular-weight components, leaving a purified sample exchanged into the selected buffer

DNA & RNA Purification & Analysis, DNA Extraction, PCR Product Clean-Up

Shipping Condition: Room Temperature