Please login to your online account to display your discounted pricing

Applied Biosystems™ Centri-Sep™ Spin Columns DFS Item

Fast and efficient purification of nucleic acids

$207.00 - $544.00

Specifications

Format Spin Column
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

401762

DFS Item
 
life technologies
LS401762
100 columns Each for $544.00

401763

DFS Item
 
life technologies
LS401763
32 columns Each for $207.00
Description & Specifications

Specifications

Format Spin Column

  • Centri-Sep™ Spin Columns use Centri-Sep™ gel that provides excellent recovery of DNA fragments larger than 16bp, while removing greater than 98% of salts, NTPs, and other low molecular weight impurities
  • Features of Centri-Sep™ columns: Processes samples faster than ultrafiltration columns
  • Achieves rapid and efficient separation
  • Permits use of custom buffers
  • Convenient 20–100μL sample size
  • Dry gel format
  • Room temperature storage
  • Microcentrifuge format
  • Designed for performance: The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Sep; unit consists of a specially fritted microfuge tube, dry gel, a wash tube, and a sample collection tube, all specifically designed for this purpose
  • Excellent recovery The Centri-Sep gel provides excellent recovery of DNA fragments with sizes greater than 16bp or a 25-mer, while removing >98% of salts, NTPs, and other unwanted low molecular weight impurities
  • Versatile The column gel is hydrated with either reagent-grade water or a buffer of your choice, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. Your sample is then applied to the column and spun again, leaving you not only with a sample purified through removal of low molecular weight components, but also exchanged into the buffer of your choice

DNA & RNA Purification & Analysis, DNA Extraction, Sequencing Reaction Purification

Shipping Condition: Room Temperature