Please login to your online account to display your discounted pricing
Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Buffer II and MgCl2 DFS Item
Chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation
Manufacturer: applied biosystems™  LSN8080243
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. N8080243
  • / Each
Description & Specifications
Specifications
Includes 6 tubes x 50μL each of AmpliTaq Gold DNA Polymerase (at 5U/μL, 250 units total per tube), 6 x 1.5mL of Buffer II (100mM Tris-HCl, pH 8.3, 500mM KCl), 6 x 25mM magnesium chloride.
Overhang 3'-A
Quantity 1500 units
For Use With (Application) Hot Start PCR, Multiplex PCR, PCR, PCR and Real-Time PCR, Routine PCR
Size (Final Product) 5kb or less