Please login to your online account to display your discounted pricing
Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Buffer I DFS Item
Chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation
Manufacturer: applied biosystems™  LSN8080242
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. N8080242
  • / Each
Description & Specifications
Specifications
Overhang 3'-A
Quantity 6 x 250 units
For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Size (Final Product) 5kb or less
Includes 6 tubes x 50μL each of AmpliTaq Gold DNA Polymerase (at 5U/μL, 250 units total per tube), 6 x 1.5mL of Buffer I (100mM Tris-HCl, pH 8.3, 500mM KCl, 15mM magnesium chloride, 0.01% (w/v) gelatin).