missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer II (250 units/tube) DFS Item

94 kDa, thermostable, recombinant DNA polymerase

$185.00 - $1,830.00

Specifications

Storage Requirements Store at -20°C.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Contains 6 x 50μL AmpliTaq DNA Polymerase (5U/μL) and 12 x 1.5mL of GeneAmp 10X PCR Buffer [100mM Tris-HCl, pH 8.3, 500mM KCl, 15mM magnesium chloride, and 0.01% (w/v) gelatin.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N8080153 Applied Biosystems™
LSN8080153
3000 units Each for $1,830.00
N8080167 Applied Biosystems™
LSN8080167
1500 units Each for $1,024.00
N8080161 Applied Biosystems™
LSN8080161
250 units Each for $185.00
Specifications

Description

AmpliTaq DNA Polymerase is obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli., and is most thoroughly characterized enzyme available for PCR process. Its recombinant nature and purification method provide unparalleled purity and reproducibility.

  • Characterized enzyme available for PCR process
  • QC-tested to guarantee reproducible results
  • Ideal for PCR

PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

Store at -20°C.