Please login to your online account to display your discounted pricing
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer II (250 units/tube) DFS Item
94 kDa, thermostable, recombinant DNA polymerase
Manufacturer: applied biosystems™  LSN8080167
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. N8080167
  • / Each
Description & Specifications
Specifications
Exonuclease Activity 5' - 3'
Fidelity 1 X
Includes 1000 units of AmpliTaq DNA Polymerase and Buffer I.
Overhang 3'-A
Polymerase AmpliTaq™ DNA Polymerase
Quantity 1500 units
Size (Final Product) 5 kb or less
Storage Requirements Store at –15 to –30°C.