missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I (250 units/tube) DFS Item

A 94 kDa, thermostable, recombinant DNA polymerase

$215.00 - $1,830.00

Specifications

Storage Requirements Store at -20°C.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Contents: 12 x 50μL each of 5U/μL of AmpliTaq DNA Polymerase; 12 x 1.5mL of GeneAmp 10X PCR Buffer (100mM Tris-HCl pH 8.3, 500mM KCl, 15mM MgCl2, 0.01% (w/v) gelatin).


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N8080152 Applied Biosystems™
LSN8080152
3000 units Each for $1,830.00
N8080160 Applied Biosystems™
LSN8080160
250 units Each for $215.00
N8080166 Applied Biosystems™
LSN8080166
1500 units Each for $1,024.00
Specifications

Description

AmpliTaq™ DNA Polymerase is recombinant DNA polymerase obtained by expression of modified form of Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is thoroughly characterized enzyme available for PCR process and its recombinant nature and purification method provide unparalleled purity and reproducibility

  • Characterized enzyme available for the PCR process
  • QC-tested to guarantee reproducible results
  • Ideal for PCR

PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

Store at -20°C.