Please login to your online account to display your discounted pricing
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I (250 units/tube) DFS Item
A 94 kDa, thermostable, recombinant DNA polymerase
Manufacturer: applied biosystems™  LSN8080166
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. N8080166
  • / Each
Description & Specifications
Specifications
Storage Requirements Store at -20°C.
Quantity 1500 units
Includes 5 x 200μL of AmpliTaq DNA Polymerase (at 5U/μL); 4 x 1.5mL of GeneAmp 10X PCR Buffer (100mM Tris-HCl pH 8.3, 500mM KCl, 15mM MgCl2,0.01% (w/v) gelatin.
Exonuclease Activity 5' - 3'
Fidelity 1 X
Polymerase AmpliTaq™ DNA Polymerase
Overhang 3'-A
Size (Final Product) 5 kb or less