Please login to your online account to display your discounted pricing

Apoptosis Activator 2

Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines

$430.93 - $430.93

Specifications

CAS 79183-19-0
Formula Weight 306.14g/mol
Synonym 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione
Storage Requirements 2° to 8°C
Format Powder
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65320LB0

 
alfa aesar™
J65320LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ65320MB

 
alfa aesar™
J65320MB
25mg Each for $430.93
Description & Specifications

Specifications

CAS 79183-19-0
Formula Weight 306.14g/mol
Synonym 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione
Storage Requirements 2° to 8°C
Format Powder
Sensitivity Light Sensitive
For Use With (Application) Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
Description Orange