MilliporeSigma™ ApopTag™ Stop/Wash Buffer
Qualified for use with Apoptag In Situ Apoptosis Detection Kits
Supplier:  MilliporeSigma™ S7108
Catalog No. S7108
Includes: Each bottle contains 20ml of ApopTag Stop/Wash Buffer
Description
Description
Intended for use in labeling the 3'-OH ends of fragmented DNA during apoptotic cell detection
Specifications
Specifications
-15°C to -25°C
Labeling the 3'-OH ends of fragmented DNA during apoptotic cell detection
20 mL