Please login to your online account to display your discounted pricing

Apigenin, Tocris


Protein kinase inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS122750

 View more versions of this product

Catalog No. 122750

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 270.2
Quantity 50mg
Molecular Formula C15H10O5
Assay Percent Range >98%
CAS 520-36-5
Synonym 5, 7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Recommended Storage Desiccate at +4°C
Chemical Name or Material Apigenin

Specifications

Chemical Name or Material Apigenin
Assay Percent Range >98%
Molecular Formula C15H10O5
Formula Weight 270.2
Synonym 5, 7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Accessories

Desiccate at +4°C